1,2,3,4-Tetrahydro-1,8-naphthyridine 95%
Leverantör: Apollo Scientific
OR926199-1G OR926199-250MG
APOSOR926199-1GEA 4425 SEK
APOSOR926199-1G APOSOR926199-250MG
1,2,3,4-Tetrahydro-1,8-naphthyridine 95%
1,2,3,4-Tetrahydro-1,8-naphthyridine
Formel: C₈H₁₀N₂
MW: 134.178 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD08234598
CAS nummer: 13623-87-5