3-Hydroxy-5-methyl-pyrazole-1-carboxylic acid tert-butyl ester 95%
Leverantör: Apollo Scientific
Synonyms: tert-Butyl 3-hydroxy-5-methyl-1H-pyrazole-1-carboxylate
OR925965-1G OR925965-250MG
APOSOR925965-1GEA 7625 SEK
APOSOR925965-1G APOSOR925965-250MG
3-Hydroxy-5-methyl-pyrazole-1-carboxylic acid tert-butyl ester 95%
3-Hydroxy-5-methyl-pyrazole-1-carboxylic acid tert-butyl ester
Formel: C₉H₁₄N₂O₃
MW: 198,219 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD22565664
CAS nummer: 915396-44-0