2-Chloro-N-(2-(hydroxymethyl)phenyl)acetamide 95%
Leverantör: Apollo Scientific
OR925723-1G OR925723-250MG
APOSOR925723-1GEA 5150 SEK
APOSOR925723-1G APOSOR925723-250MG
2-Chloro-N-(2-(hydroxymethyl)phenyl)acetamide 95%
2-Chloro-N-(2-(hydroxymethyl)phenyl)acetamide
Formel: C₉H₁₀ClNO₂
MW: 199,634 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD12124629
CAS nummer: 189940-09-8