5,7-Dihydroxycoumarin 95%
Leverantör: Apollo Scientific
Synonyms: 5,7-Dihydroxy-2H-chromen-2-one
OR925694-1G OR925694-250MG OR925694-5G
APOSOR925694-1GEA 1200 SEK
APOSOR925694-1G APOSOR925694-250MG APOSOR925694-5G
5,7-Dihydroxycoumarin 95%
5,7-Dihydroxycoumarin
Formel: C₉H₆O₄
MW: 178,142 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00488354
CAS nummer: 2732-18-5