Methyl 2-methoxy-5-methylbenzoate 95%
Leverantör: Apollo Scientific
OR924992-1G OR924992-5G
APOSOR924992-1GEA 290 SEK
APOSOR924992-1G APOSOR924992-5G
Methyl 2-methoxy-5-methylbenzoate 95%
Methyl 2-methoxy-5-methylbenzoate
Formel: C₁₀H₁₂O₃
Storage Temperature: Omgivande
MDL Number: MFCD06411343
CAS nummer: 63113-79-1