Ethyl-3-isopropyl pyrazole-4-carboxylate 95%
Leverantör: Apollo Scientific
Synonyms: Ethyl 3-isopropyl-1H-pyrazole-4-carboxylate
OR924858-1G OR924858-250MG
APOSOR924858-1GEA 5750 SEK
APOSOR924858-1G APOSOR924858-250MG
Ethyl-3-isopropyl pyrazole-4-carboxylate 95%
Ethyl-3-isopropyl pyrazole-4-carboxylate
Formel: C₉H₁₄N₂O₂
MW: 182,22 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD07369096
CAS nummer: 342026-17-9