(S)-3-Methoxypiperidine hydrochloride 95%

Leverantör: Apollo Scientific

OR924852-1G OR924852-5G
APOSOR924852-1GEA 3175 SEK
APOSOR924852-1G APOSOR924852-5G
(S)-3-Methoxypiperidine hydrochloride 95%
(S)-3-Methoxypiperidine hydrochloride
Formel: C₆H₁₃NO·ClH
Storage Temperature: Omgivande
MDL Number: MFCD17015900
CAS nummer: 688809-96-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR