(R)-Methyl tetrahydrofuran-3-carboxylate 95%
Leverantör: Apollo Scientific
OR924685-1G OR924685-250MG
APOSOR924685-1GEA 9500 SEK
APOSOR924685-1G APOSOR924685-250MG
(R)-Methyl tetrahydrofuran-3-carboxylate 95%
(R)-Methyl tetrahydrofuran-3-carboxylate
Formel: C₆H₁₀O₃
MW: 130.142 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD17015960
CAS nummer: 191347-93-0