1H-Pyrrolo[2,3-b]pyridin-2-ylmethanol 95%

Leverantör: Apollo Scientific

OR923262-1G OR923262-250MG
APOSOR923262-1GEA 9500 SEK
APOSOR923262-1G APOSOR923262-250MG
1H-Pyrrolo[2,3-b]pyridin-2-ylmethanol 95%
1H-Pyrrolo[2,3-b]pyridin-2-ylmethanol
Formel: C₈H₈N₂O
MW: 148,162 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD18457923
CAS nummer: 1269479-13-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR