(S)-3-Cyanopiperidine hydrochloride 97%
Leverantör: Apollo Scientific
Varning
OR922852-1G OR922852-250MG
APOSOR922852-1GEA 9500 SEK
APOSOR922852-1G APOSOR922852-250MG
(S)-3-Cyanopiperidine hydrochloride 97%
(S)-3-Cyanopiperidine hydrochloride
Formel: C₆H₁₀N₂·ClH
MW: 146,618 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD23106073
CAS nummer: 915226-72-1