Spiro[3.3]heptan-2-one
Leverantör: Apollo Scientific
OR922744-1G OR922744-5G OR922744-250MG
APOSOR922744-1GEA 7200 SEK
APOSOR922744-1G APOSOR922744-5G APOSOR922744-250MG
Spiro[3.3]heptan-2-one
Spiro[3.3]heptan-2-one
Formel: C₇H₁₀O
MW: 110,154 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD22421860
CAS nummer: 30152-57-9