2-Methyl-5-[(methylamino)methyl]pyridine 95%
Leverantör: Apollo Scientific
OR922580-1G OR922580-250MG
APOSOR922580-1GEA 7450 SEK
APOSOR922580-1G APOSOR922580-250MG
2-Methyl-5-[(methylamino)methyl]pyridine 95%
2-Methyl-5-[(methylamino)methyl]pyridine
Formel: C₈H₁₂N₂
MW: 136,194 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD11212404
CAS nummer: 120740-02-5