2,7-Diazaspiro[3.5]nonan-1-one 97%
Leverantör: Apollo Scientific
Varning
OR922146-100MG OR922146-250MG
APOSOR922146-100MGEA 1175 SEK
APOSOR922146-100MG APOSOR922146-250MG
2,7-Diazaspiro[3.5]nonan-1-one 97%
2,7-Diazaspiro[3.5]nonan-1-one
Formel: C₇H₁₂N₂O
MW: 140,183 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD13180578
CAS nummer: 1147422-92-1