3-Chloro-4,6-dimethylpyridazine
Leverantör: Apollo Scientific
OR921631-1G OR921631-5G
APOSOR921631-1GEA 6950 SEK
APOSOR921631-1G APOSOR921631-5G
3-Chloro-4,6-dimethylpyridazine
3-Chloro-4,6-dimethylpyridazine
Formel: C₆H₇ClN₂
MW: 142,586 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD17016067
CAS nummer: 17258-26-3