4,5-Dichlorothiophene-2-carboxylic acid 98%
Leverantör: Apollo Scientific
OR920479-1G OR920479-250MG OR920479-5G
APOSOR920479-1GEA 825 SEK
APOSOR920479-1G APOSOR920479-250MG APOSOR920479-5G
4,5-Dichlorothiophene-2-carboxylic acid 98%
4,5-Dichlorothiophene-2-carboxylic acid
Formel: C₅H₂Cl₂O₂S
MW: 197,039 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD07366674
CAS nummer: 31166-29-7