6,7-Dihydro-1,7-naphthyridin-8(5H)-one 98%
Leverantör: Apollo Scientific
OR919197-1G OR919197-250MG
APOSOR919197-1GEA 8050 SEK
APOSOR919197-1G APOSOR919197-250MG
6,7-Dihydro-1,7-naphthyridin-8(5H)-one 98%
6,7-Dihydro-1,7-naphthyridin-8(5H)-one
Formel: C₈H₈N₂O
MW: 148,162 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD16658787
CAS nummer: 301666-63-7