4,5-Dichloro-2-methylpyrimidine 98%
Leverantör: Apollo Scientific
OR918935-1G OR918935-250MG
APOSOR918935-1GEA 1475 SEK
APOSOR918935-1G APOSOR918935-250MG
4,5-Dichloro-2-methylpyrimidine 98%
4,5-Dichloro-2-methylpyrimidine
Formel: C₅H₄Cl₂N₂
MW: 163,005 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD16987924
CAS nummer: 26740-71-6