2-Methyl-3H-thieno[3,2-d]pyrimidin-4-one 95%
Leverantör: Apollo Scientific
Synonyms: 2-Methylthieno[3,2-d]pyrimidin-4(3H)-one
OR918771-1G OR918771-5G
APOSOR918771-1GEA 1075 SEK
APOSOR918771-1G APOSOR918771-5G
2-Methyl-3H-thieno[3,2-d]pyrimidin-4-one 95%
2-Methyl-3H-thieno[3,2-d]pyrimidin-4-one
Formel: C₇H₆N₂OS
MW: 166,2 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09863154
CAS nummer: 18678-13-2