3,4-Dihydro-2H-benzo[b]oxepin-5-one 95%

Leverantör: Apollo Scientific

OR918393-1G OR918393-250MG
APOSOR918393-1GEA 8400 SEK
APOSOR918393-1G APOSOR918393-250MG
3,4-Dihydro-2H-benzo[b]oxepin-5-one 95%
3,4-Dihydro-2H-benzo[b]oxepin-5-one
Formel: C₁₀H₁₀O₂
MW: 162,185 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD01648756
CAS nummer: 6786-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR