4-(1-Hydroxy-1-methylethyl)benzoic acid 97%
Leverantör: Apollo Scientific
OR917681-1G OR917681-250MG
APOSOR917681-1GEA 8375 SEK
APOSOR917681-1G APOSOR917681-250MG
4-(1-Hydroxy-1-methylethyl)benzoic acid 97%
4-(1-Hydroxy-1-methylethyl)benzoic acid
Formel: C₁₀H₁₂O₃
MW: 180.2 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD02185844
CAS nummer: 3609-50-5