6,7-Dihydro-5h-cyclopenta[b]pyridin-7-amine 95%
Leverantör: Apollo Scientific
OR917333-100MG
OR917333-250MG
APOSOR917333-100MGEA
5075
SEK
APOSOR917333-100MG
APOSOR917333-250MG
6,7-Dihydro-5h-cyclopenta[b]pyridin-7-amine 95%
6,7-Dihydro-5h-cyclopenta[b]pyridin-7-amine
Formel:
C₈H₁₀N₂ MW: 134,178 g/mol Storage Temperature: Kylskåp |
MDL Number:
MFCD07778343 CAS nummer: 185122-75-2 UN: 3259 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...