4-Chloro-2,3-dihydropyridazin-3-one
Leverantör: Apollo Scientific
Synonyms: 4-Chloropyridazin-3-ol
OR916662-10G OR916662-1G OR916662-5G
APOSOR916662-10GEA 7750 SEK
APOSOR916662-10G APOSOR916662-1G APOSOR916662-5G
4-Chloro-2,3-dihydropyridazin-3-one
4-Chloro-2,3-dihydropyridazin-3-one
Formel: C₄H₃ClN₂O
MW: 130,532 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD08062119
CAS nummer: 1677-79-8