2-Chlorooxazolo[5,4-b]pyridine 97%
Leverantör: Apollo Scientific
Varning
OR916520-250MG OR916520-1G
APOSOR916520-250MGEA 2500 SEK
APOSOR916520-250MG APOSOR916520-1G
2-Chlorooxazolo[5,4-b]pyridine 97%
2-Chlorooxazolo[5,4-b]pyridine
Formel: C₆H₃ClN₂O
MW: 154,554 g/mol
Storage Temperature: Frys
MDL Number: MFCD18802457
CAS nummer: 159870-95-8