1,1-Dioxo-isothiazolidine-3-carboxylic acid 97%
Leverantör: Apollo Scientific
Varning
OR914154-1G OR914154-5G
APOSOR914154-1GEA 7950 SEK
APOSOR914154-1G APOSOR914154-5G
1,1-Dioxo-isothiazolidine-3-carboxylic acid 97%
1,1-Dioxo-isothiazolidine-3-carboxylic acid
Formel: C₄H₇NO₄S
Storage Temperature: Omgivande
MDL Number: MFCD07369945
CAS nummer: 1146957-01-8