(1-Methyl-5-phenyl-1H-pyrazol-3-yl)methylamine 95+%
Leverantör: Apollo Scientific
Fara
Synonyms: [3-(Aminomethyl)-1-methyl-1H-pyrazol-5-yl]benzene, 3-(Aminomethyl)-1-methyl-5-phenyl-1H-pyrazole
OR9131-1G OR9131-250MG
APOSOR9131-1GEA 3700 SEK
APOSOR9131-1G APOSOR9131-250MG
(1-Methyl-5-phenyl-1H-pyrazol-3-yl)methylamine 95+%
(1-Methyl-5-phenyl-1H-pyrazol-3-yl)methylamine
Formel: C₁₁H₁₃N₃
MW: 187,241 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD08271937
CAS nummer: 869901-12-2
UN: 2735
ADR: 8,III