N-Boc-2-aminoacetaldehyde 95%

Leverantör: Apollo Scientific

OR912671-1G OR912671-5G
APOSOR912671-1GEA 1575 SEK
APOSOR912671-1G APOSOR912671-5G
N-Boc-2-aminoacetaldehyde 95%
N-Boc-2-aminoacetaldehyde
Formel: C₇H₁₃NO₃
MW: 159,183 g/mol
Storage Temperature: Frys
MDL Number: MFCD01321273
CAS nummer: 89711-08-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR