(1,4-Dioxa-spiro[4.4]non-6-yl)-methanol 90%
Leverantör: Apollo Scientific
OR912514-1G OR912514-250MG
APOSOR912514-1GEA 8050 SEK
APOSOR912514-1G APOSOR912514-250MG
(1,4-Dioxa-spiro[4.4]non-6-yl)-methanol 90%
(1,4-Dioxa-spiro[4.4]non-6-yl)-methanol
Formel: C₈H₁₄O₃
MW: 158.195 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09991662
CAS nummer: 23153-74-4