2-Ethoxy-3-nitrotoluene 98%
Leverantör: Apollo Scientific
OR911220-1G OR911220-5G
APOSOR911220-1GEA 1975 SEK
APOSOR911220-1G APOSOR911220-5G
2-Ethoxy-3-nitrotoluene 98%
2-Ethoxy-3-nitrotoluene
Formel: C₉H₁₁NO₃
MW: 181.189 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD12547885
CAS nummer: 1208074-91-2