1-(2-Chlorophenyl)ethanamine hydrochloride 98%
Leverantör: Apollo Scientific
OR910988-1G OR910988-5G
APOSOR910988-1GEA 2575 SEK
APOSOR910988-1G APOSOR910988-5G
1-(2-Chlorophenyl)ethanamine hydrochloride 98%
1-(2-Chlorophenyl)ethanamine hydrochloride
Formel: C₈H₁₀ClN·ClH
MW: 192.086 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD11841333
CAS nummer: 856629-37-3