4-Cyanogramine 95%
Leverantör: Apollo Scientific
OR910602-1G
OR910602-5G
APOSOR910602-1GEA
2925
SEK
APOSOR910602-1G
APOSOR910602-5G
4-Cyanogramine 95%
4-Cyanogramine
Formel:
C₁₂H₁₃N₃ Storage Temperature: Omgivande |
MDL Number:
MFCD11112015 CAS nummer: 105907-63-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...