6-Cyanogramine 98%
Leverantör: Apollo Scientific
OR910600-1G
OR910600-5G
APOSOR910600-1GEA
2925
SEK
APOSOR910600-1G
APOSOR910600-5G
6-Cyanogramine 98%
6-Cyanogramine
Formel:
C₁₂H₁₃N₃ Storage Temperature: Omgivande |
MDL Number:
MFCD11112020 CAS nummer: 379234-99-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...