4-(1H-Pyrazol-4-yl)benzonitrile 95%
Leverantör: Apollo Scientific
OR910287-1G OR910287-250MG
APOSOR910287-1GEA 9500 SEK
APOSOR910287-1G APOSOR910287-250MG
4-(1H-Pyrazol-4-yl)benzonitrile 95%
4-(1H-Pyrazol-4-yl)benzonitrile
Formel: C₁₀H₇N₃
MW: 169,183 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD11933267
CAS nummer: 1101167-56-9