2-Chloro-N,N-dimethylpyrimidin-4-amine 98%
Leverantör: Apollo Scientific
OR909499-1G OR909499-5G
APOSOR909499-1GEA 2575 SEK
APOSOR909499-1G APOSOR909499-5G
2-Chloro-N,N-dimethylpyrimidin-4-amine 98%
2-Chloro-N,N-dimethylpyrimidin-4-amine
Formel: C₆H₈ClN₃
MW: 157.601 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00129722
CAS nummer: 31058-81-8