2-Ethoxy-3-isopropoxypyridine 98%
Leverantör: Apollo Scientific
OR909397-1G OR909397-25G OR909397-5G
APOSOR909397-1GEA 1475 SEK
APOSOR909397-1G APOSOR909397-25G APOSOR909397-5G
2-Ethoxy-3-isopropoxypyridine 98%
2-Ethoxy-3-isopropoxypyridine
Formel: C₁₀H₁₅NO₂
Storage Temperature: Omgivande
MDL Number: MFCD19981544
CAS nummer: 1330750-22-5