(3-Formyloxyphenyl)boronic acid 95%

Leverantör: Apollo Scientific

OR908506-1G OR908506-5G
APOSOR908506-1GEA 1300 SEK
APOSOR908506-1G APOSOR908506-5G
(3-Formyloxyphenyl)boronic acid 95%
(3-Formyloxyphenyl)boronic acid
Formel: C₇H₇BO₄
MW: 165,939 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD16659866
CAS nummer: 1309981-04-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR