Methyl 5-methoxy-2-methylbenzoate 98%
Leverantör: Apollo Scientific
OR908053-10G OR908053-1G OR908053-5G
APOSOR908053-10GEA 8925 SEK
APOSOR908053-10G APOSOR908053-1G APOSOR908053-5G
Methyl 5-methoxy-2-methylbenzoate 98%
Methyl 5-methoxy-2-methylbenzoate
Formel: C₁₀H₁₂O₃
MW: 180,2 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD11045607
CAS nummer: 73502-03-1