4-Bromo-3-hydroxybenzaldehyde 95+%
Leverantör: Apollo Scientific
Varning
OR907915-1G OR907915-250MG
APOSOR907915-1GEA 1125 SEK
APOSOR907915-1G APOSOR907915-250MG
4-Bromo-3-hydroxybenzaldehyde 95+%
4-Bromo-3-hydroxybenzaldehyde
Formel: C₇H₅BrO₂
MW: 201.017 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD11100992
CAS nummer: 20035-32-9