2-(1-Methylpiperidin-4-ylidene)acetonitrile 97%
Leverantör: Apollo Scientific
OR907453-10G
OR907453-1G
OR907453-5G
APOSOR907453-10GEA
5900
SEK
APOSOR907453-10G
APOSOR907453-1G
APOSOR907453-5G
2-(1-Methylpiperidin-4-ylidene)acetonitrile 97%
2-(1-Methylpiperidin-4-ylidene)acetonitrile
Formel:
C₈H₁₂N₂ MW: 136,194 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD13178818 CAS nummer: 37123-91-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...