2-Pyridin-4-yl-azepane 95%
Leverantör: Apollo Scientific
OR906618-1G
OR906618-250MG
APOSOR906618-1GEA
8050
SEK
APOSOR906618-1G
APOSOR906618-250MG
2-Pyridin-4-yl-azepane 95%
2-Pyridin-4-yl-azepane
Formel:
C₁₁H₁₆N₂ MW: 176,258 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD02663701 CAS nummer: 383129-02-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...