2-(3-Methylbenzyl)pyrrolidine 95%
Leverantör: Apollo Scientific
OR906476-1G
OR906476-5G
APOSOR906476-1GEA
2575
SEK
APOSOR906476-1G
APOSOR906476-5G
2-(3-Methylbenzyl)pyrrolidine 95%
2-(3-Methylbenzyl)pyrrolidine
Formel:
C₁₂H₁₇N MW: 175,27 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD02663495 CAS nummer: 383127-61-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...