2-(4-Methoxy-3-methylphenyl)pyrrolidine 95%
Leverantör: Apollo Scientific
OR906383-1G
OR906383-250MG
APOSOR906383-1GEA
10175
SEK
APOSOR906383-1G
APOSOR906383-250MG
2-(4-Methoxy-3-methylphenyl)pyrrolidine 95%
2-(4-Methoxy-3-methylphenyl)pyrrolidine
Formel:
C₁₂H₁₇NO MW: 191,269 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD05189324 CAS nummer: 887361-09-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...