5-tert-Butyl-1H-pyrazole-3-carboxylic acid 97%
Leverantör: Apollo Scientific
OR905580-1G OR905580-5G
APOSOR905580-1GEA 3175 SEK
APOSOR905580-1G APOSOR905580-5G
5-tert-Butyl-1H-pyrazole-3-carboxylic acid 97%
5-tert-Butyl-1H-pyrazole-3-carboxylic acid
Formel: C₈H₁₂N₂O₂
MW: 168,193 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00817563
CAS nummer: 83405-71-4