Ethyl 5-ethyl-1H-pyrazole-3-carboxylate 97%
Leverantör: Apollo Scientific
OR905371-1G OR905371-5G
APOSOR905371-1GEA 2250 SEK
APOSOR905371-1G APOSOR905371-5G
Ethyl 5-ethyl-1H-pyrazole-3-carboxylate 97%
Ethyl 5-ethyl-1H-pyrazole-3-carboxylate
Formel: C₈H₁₂N₂O₂
MW: 168.193 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD08277223
CAS nummer: 26308-40-7