2-(2-Furyl)piperidine 95%
Leverantör: Apollo Scientific
OR904721-1G
OR904721-250MG
APOSOR904721-1GEA
10175
SEK
APOSOR904721-1G
APOSOR904721-250MG
2-(2-Furyl)piperidine 95%
2-(2-Furyl)piperidine
Formel:
C₉H₁₃NO MW: 151,206 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD02663560 CAS nummer: 97073-24-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...