4-Ethylphenyl isocyanide 95%

Leverantör: Apollo Scientific

OR904368-1G OR904368-5G
APOSOR904368-1GEA 2425 SEK
APOSOR904368-1G APOSOR904368-5G
4-Ethylphenyl isocyanide 95%
4-Ethylphenyl isocyanide
Formel: C₉H₉N
MW: 131,174 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00972858
CAS nummer: 143063-89-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR