2,4-Dimethyl-6-(2-pyrrolidinylmethyl)pyridine 95%
Leverantör: Apollo Scientific
OR902810-1G
OR902810-250MG
APOSOR902810-1GEA
9500
SEK
APOSOR902810-1G
APOSOR902810-250MG
2,4-Dimethyl-6-(2-pyrrolidinylmethyl)pyridine 95%
2,4-Dimethyl-6-(2-pyrrolidinylmethyl)pyridine
Formel:
C₁₂H₁₈N₂ MW: 190,285 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD05189306 CAS nummer: 524674-46-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...