2-(4-Methylbenzyl)pyrrolidine 95%

Leverantör: Apollo Scientific

OR902761-1G OR902761-250MG
APOSOR902761-1GEA 8050 SEK
APOSOR902761-1G APOSOR902761-250MG
2-(4-Methylbenzyl)pyrrolidine 95%
2-(4-Methylbenzyl)pyrrolidine
Formel: C₁₂H₁₇N
MW: 175,27 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD02663496
CAS nummer: 383127-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR