6-Methoxynicotinamide 98%

Leverantör: Apollo Scientific

OR902093-1G OR902093-5G
APOSOR902093-1GEA 1700 SEK
APOSOR902093-1G APOSOR902093-5G
6-Methoxynicotinamide 98%
6-Methoxynicotinamide
Formel: C₇H₈N₂O₂
Storage Temperature: Omgivande
MDL Number: MFCD00229166
CAS nummer: 7150-23-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR