2,3-Dichloro-5-methoxypyridine 98%
Leverantör: Apollo Scientific
OR901895-1G OR901895-5G
APOSOR901895-1GEA 1975 SEK
APOSOR901895-1G APOSOR901895-5G
2,3-Dichloro-5-methoxypyridine 98%
2,3-Dichloro-5-methoxypyridine
Formel: C₆H₅Cl₂NO
MW: 178,016 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD08277295
CAS nummer: 885168-12-7