2-Propoxyquinoline 95%

Leverantör: Apollo Scientific

OR901817-1G OR901817-5G
APOSOR901817-1GEA 1725 SEK
APOSOR901817-1G APOSOR901817-5G
2-Propoxyquinoline 95%
2-Propoxyquinoline
Formel: C₁₂H₁₃NO
Storage Temperature: Omgivande
MDL Number: MFCD18783197
CAS nummer: 945-83-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR